Product Details

SNP ID
rs145208358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53285009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGAGAGTCCATTAAAAGACACC[A/G]CATTAACTACAACAAAAAGAAGAGC
Phenotype
MIM: 607010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCP1A PubMed Links

Gene Details

Gene
DCP1A
Gene Name
decapping mRNA 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290204.1 4316 UTR 3 NP_001277133.1
NM_001290205.1 4316 UTR 3 NP_001277134.1
NM_001290206.1 4316 UTR 3 NP_001277135.1
NM_001290207.1 4316 UTR 3 NP_001277136.1
NM_018403.6 4316 UTR 3 NP_060873.4

View Full Product Details