Product Details

SNP ID
rs145299223
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53865275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGTGTCAGCAGGAAAAAGATC[A/G]CAAGCCTGCCACGATGGCTGCTGCT
Phenotype
MIM: 605458
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACTR8 PubMed Links

Gene Details

Gene
ACTR8
Gene Name
ARP8 actin-related protein 8 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022899.4 1384 Intron NP_075050.3
XM_005265587.4 1384 Intron XP_005265644.1
XM_011534249.2 1384 Intron XP_011532551.1
Gene
IL17RB
Gene Name
interleukin 17 receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018725.3 1384 Silent Mutation TCA,TCG S492S NP_061195.2
XM_005265310.4 1384 Silent Mutation TCA,TCG S521S XP_005265367.1
XM_005265311.4 1384 Silent Mutation TCA,TCG S505S XP_005265368.1
XM_005265312.4 1384 Silent Mutation TCA,TCG S476S XP_005265369.1
XM_011533941.2 1384 UTR 3 XP_011532243.1
XM_017006804.1 1384 Silent Mutation TCA,TCG S404S XP_016862293.1
XM_017006805.1 1384 Silent Mutation TCA,TCG S388S XP_016862294.1
XM_017006806.1 1384 Silent Mutation TCA,TCG S375S XP_016862295.1
XM_017006807.1 1384 UTR 3 XP_016862296.1

View Full Product Details