Product Details

SNP ID
rs145384643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:159273850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGTCCGGGCAGCGTGTTACAAC[A/G]TGGGACTGTGACCAGGGCAAGCACT
Phenotype
MIM: 611622
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IQCJ PubMed Links

Gene Details

Gene
IQCJ
Gene Name
IQ motif containing J
There are no transcripts associated with this gene.

Gene
IQCJ-SCHIP1
Gene Name
IQCJ-SCHIP1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197113.1 604 Intron NP_001184042.1
NM_001197114.1 604 Intron NP_001184043.1
Gene
MIR3919
Gene Name
microRNA 3919
There are no transcripts associated with this gene.

Gene
SCHIP1
Gene Name
schwannomin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197107.1 604 Silent Mutation ACA,ACG T10T NP_001184036.1
NM_001197108.1 604 Silent Mutation ACA,ACG T10T NP_001184037.1
NM_001197109.1 604 Intron NP_001184038.1
NM_014575.3 604 Silent Mutation ACA,ACG T10T NP_055390.1

View Full Product Details