Product Details

SNP ID
rs145394428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49016485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTGATGGTGGCCACAGAGAGAA[C/T]ACCAAAGATCTCTGTCCAGGCTGGG
Phenotype
MIM: 146691 MIM: 615150 MIM: 612911 MIM: 614584 MIM: 606031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DALRD3 PubMed Links

Gene Details

Gene
DALRD3
Gene Name
DALR anticodon binding domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009996.2 1466 Missense Mutation ATT,GTT I364V NP_001009996.1
NM_001276405.1 1466 Missense Mutation ATT,GTT I364V NP_001263334.1
NM_018114.5 1466 Missense Mutation ATT,GTT I197V NP_060584.3
XM_005265269.3 1466 Missense Mutation ATT,GTT I364V XP_005265326.1
XM_006713219.2 1466 Missense Mutation ATT,GTT I197V XP_006713282.1
XM_011533891.1 1466 Missense Mutation ATT,GTT I364V XP_011532193.1
XM_011533892.1 1466 Missense Mutation ATT,GTT I197V XP_011532194.1
XM_011533893.1 1466 Missense Mutation TAT,TGT Y343C XP_011532195.1
XM_017006723.1 1466 Missense Mutation ATT,GTT I197V XP_016862212.1
Gene
IMPDH2
Gene Name
inosine monophosphate dehydrogenase 2
There are no transcripts associated with this gene.

Gene
MIR191
Gene Name
microRNA 191
There are no transcripts associated with this gene.

Gene
MIR425
Gene Name
microRNA 425
There are no transcripts associated with this gene.

Gene
NDUFAF3
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
There are no transcripts associated with this gene.

Gene
P4HTM
Gene Name
prolyl 4-hydroxylase, transmembrane
There are no transcripts associated with this gene.

Gene
WDR6
Gene Name
WD repeat domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320546.1 1466 Intron NP_001307475.1
NM_001320547.1 1466 Intron NP_001307476.1
NM_018031.4 1466 Intron NP_060501.3

View Full Product Details