Product Details

SNP ID
rs145542585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98354561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATCCAAGGAGGGAAGAGGTAA[A/G]GCATTTTCTACCTGTGCATCCCACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5K4 PubMed Links

Gene Details

Gene
OR5K4
Gene Name
olfactory receptor family 5 subfamily K member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005517.1 708 Silent Mutation AAA,AAG K236K NP_001005517.1

View Full Product Details