Product Details

SNP ID
rs145610267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:147410298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGCTGGCTTCGGCCGGCCAGAC[A/G]GCCTTCACGTCGCAGGCGCCAGGCT
Phenotype
MIM: 600470 MIM: 608948
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZIC1 PubMed Links

Gene Details

Gene
ZIC1
Gene Name
Zic family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003412.3 905 Silent Mutation ACA,ACG T62T NP_003403.2
Gene
ZIC4
Gene Name
Zic family member 4
There are no transcripts associated with this gene.

View Full Product Details