Product Details

SNP ID
rs145856089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:139456144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGACCCTGATGCTAGCTGATTT[G/T]CTTCCAAGTATGTGCAAACTACATT
Phenotype
MIM: 180280
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100507291 PubMed Links

Gene Details

Gene
LOC100507291
Gene Name
uncharacterized LOC100507291
There are no transcripts associated with this gene.

Gene
RBP2
Gene Name
retinol binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004164.2 Intron NP_004155.2

View Full Product Details