Product Details
- SNP ID
-
rs145875310
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:113954192 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGAGTGCCTGAGCAGAAAAGGGCA[A/G]GAGAAGACCAAAGGGTTCCCTGGGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC191
PubMed Links
Gene Details
- Gene
- CCDC191
- Gene Name
- coiled-coil domain containing 191
There are no transcripts associated with this gene.
- Gene
- GRAMD1C
- Gene Name
- GRAM domain containing 1C
There are no transcripts associated with this gene.
- Gene
- ZDHHC23
- Gene Name
- zinc finger DHHC-type containing 23
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001320466.1 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
NP_001307395.1 |
NM_001320467.1 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
NP_001307396.1 |
NM_001320468.1 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
NP_001307397.1 |
NM_173570.4 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
NP_775841.2 |
XM_005247269.1 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
XP_005247326.1 |
XM_005247270.3 |
876 |
Silent Mutation |
CAA,CAG |
Q57Q |
XP_005247327.1 |
XM_006713562.3 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
XP_006713625.1 |
XM_011512618.2 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
XP_011510920.1 |
XM_011512619.1 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
XP_011510921.1 |
XM_017006084.1 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
XP_016861573.1 |
XM_017006085.1 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
XP_016861574.1 |
XM_017006086.1 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
XP_016861575.1 |
XM_017006087.1 |
876 |
Silent Mutation |
CAA,CAG |
Q218Q |
XP_016861576.1 |
View Full Product Details