Product Details

SNP ID
rs145949970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44594506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGAGAATCCACACTGGACTGAAG[C/G]CCTATACATGCAGTGAATGTGGGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF660 PubMed Links

Gene Details

Gene
ZNF660
Gene Name
zinc finger protein 660
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173658.2 646 Missense Mutation CCC,GCC P105A NP_775929.2
XM_005265090.4 646 Missense Mutation CCC,GCC P105A XP_005265147.1
XM_006713117.3 646 Missense Mutation CCC,GCC P105A XP_006713180.1
XM_011533622.2 646 Missense Mutation CCC,GCC P105A XP_011531924.1

View Full Product Details