Product Details

SNP ID
rs146270961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:15450364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACCAGGCCCCAAGCTCCAGCCT[A/G]CTGAGTCCCCAAGACACAGGCTCAT
Phenotype
MIM: 603033 MIM: 608315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COLQ PubMed Links

Gene Details

Gene
COLQ
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005677.3 2903 UTR 3 NP_005668.2
NM_080538.2 2903 UTR 3 NP_536799.1
NM_080539.3 2903 UTR 3 NP_536800.2
Gene
EAF1
Gene Name
ELL associated factor 1
There are no transcripts associated with this gene.

View Full Product Details