Product Details

SNP ID
rs146324247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122690022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTCTCTACCCAATTCCACATA[C/T]CCTTCAAGACCTGCTGAACTTCCTC
Phenotype
MIM: 610028
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP14 PubMed Links

Gene Details

Gene
PARP14
Gene Name
poly(ADP-ribose) polymerase family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017554.2 Intron NP_060024.2
XM_011512928.2 Intron XP_011511230.1
XM_011512929.2 Intron XP_011511231.1

View Full Product Details