Product Details

SNP ID
rs146411700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169778822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTATCTCAAAGTGGAAGAGGT[A/G]GTCACTAAATGCAAAATAAAGATGG
Phenotype
MIM: 608534 MIM: 606042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTRT3 PubMed Links

Gene Details

Gene
ACTRT3
Gene Name
actin related protein T3
There are no transcripts associated with this gene.

Gene
MYNN
Gene Name
myoneurin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185118.1 1508 Silent Mutation GTA,GTG V107V NP_001172047.1
NM_001185119.1 1508 Silent Mutation GTA,GTG V107V NP_001172048.1
NM_018657.4 1508 Silent Mutation GTA,GTG V107V NP_061127.1
XM_005247621.4 1508 Silent Mutation GTA,GTG V107V XP_005247678.1
XM_005247622.3 1508 Intron XP_005247679.1
XM_005247624.3 1508 UTR 5 XP_005247681.1
XM_011512987.1 1508 Silent Mutation GTA,GTG V107V XP_011511289.1
XM_017006864.1 1508 Silent Mutation GTA,GTG V107V XP_016862353.1
XM_017006865.1 1508 UTR 5 XP_016862354.1
XM_017006866.1 1508 UTR 5 XP_016862355.1
XM_017006867.1 1508 Intron XP_016862356.1
XM_017006868.1 1508 Intron XP_016862357.1

View Full Product Details