Product Details

SNP ID
rs146413045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121993241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAGGCGGCTTCTCCTCGGGCCAG[C/T]GTGGGGAGTGCGAGCCGCGGCGGTG
Phenotype
MIM: 609739
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ILDR1 PubMed Links

Gene Details

Gene
ILDR1
Gene Name
immunoglobulin like domain containing receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199799.1 1453 Missense Mutation CAC,CGC H503R NP_001186728.1
NM_001199800.1 1453 Missense Mutation CAC,CGC H414R NP_001186729.1
NM_175924.3 1453 Missense Mutation CAC,CGC H459R NP_787120.1
XM_005247389.4 1453 Missense Mutation CAC,CGC H471R XP_005247446.1
XM_011512738.2 1453 Missense Mutation CAC,CGC H503R XP_011511040.1
XM_011512739.2 1453 Missense Mutation CAC,CGC H324R XP_011511041.1

View Full Product Details