Product Details

SNP ID
rs146488757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49727293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTTGGGCTGAGAGGAGGGACC[C/T]GCCTCGGGGCTGGTGTTGCTGGGGC
Phenotype
MIM: 615691 MIM: 615320 MIM: 606991 MIM: 614472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMIGO3 PubMed Links

Gene Details

Gene
AMIGO3
Gene Name
adhesion molecule with Ig like domain 3
There are no transcripts associated with this gene.

Gene
GMPPB
Gene Name
GDP-mannose pyrophosphorylase B
There are no transcripts associated with this gene.

Gene
IP6K1
Gene Name
inositol hexakisphosphate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006115.2 1808 Silent Mutation GCA,GCG A220A NP_001006115.1
NM_001242829.1 1808 Silent Mutation GCA,GCG A385A NP_001229758.1
NM_153273.3 1808 Silent Mutation GCA,GCG A385A NP_695005.1
Gene
RNF123
Gene Name
ring finger protein 123
There are no transcripts associated with this gene.

View Full Product Details