Product Details
- SNP ID
-
rs146536201
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:9366953 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATAACTTATTTGTGGTGGTTCAG[C/G]AGTTTCAAGATTACCAGTTCAAACA
- Phenotype
-
MIM: 606525
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SRGAP3
PubMed Links
Gene Details
- Gene
- SRGAP3
- Gene Name
- SLIT-ROBO Rho GTPase activating protein 3
There are no transcripts associated with this gene.
- Gene
- THUMPD3
- Gene Name
- THUMP domain containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001114092.1 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
NP_001107564.1 |
NM_015453.2 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
NP_056268.2 |
XM_005265021.4 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_005265078.1 |
XM_005265022.4 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_005265079.1 |
XM_005265023.3 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_005265080.1 |
XM_005265024.4 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_005265081.1 |
XM_006713086.3 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_006713149.1 |
XM_011533570.2 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_011531872.1 |
XM_011533571.2 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_011531873.1 |
XM_017006113.1 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_016861602.1 |
XM_017006114.1 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_016861603.1 |
XM_017006115.1 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_016861604.1 |
XM_017006116.1 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_016861605.1 |
XM_017006117.1 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_016861606.1 |
XM_017006118.1 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_016861607.1 |
XM_017006119.1 |
1008 |
Missense Mutation |
CAG,GAG |
Q100E |
XP_016861608.1 |
View Full Product Details