Product Details

SNP ID
rs146685432
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112923724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACTTCACTTTGTAATGCCTCAG[A/T]TGCCTTCACCTTGTTTGTAGTATCA
Phenotype
MIM: 607546
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
CD200R1 PubMed Links
Additional Information
For this assay, SNP(s) [rs79556290,rs9865242] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CD200R1
Gene Name
CD200 receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138806.3 1233 Missense Mutation ACT,TCT T334S NP_620161.1
NM_138939.2 1233 Intron NP_620385.1
NM_138940.2 1233 Intron NP_620386.1
NM_170780.2 1233 Missense Mutation ACT,TCT T311S NP_740750.1

View Full Product Details