Product Details

SNP ID
rs146711138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:134601451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGGCAGCCAGCGCGGGGTCCAG[C/T]GAGTGACATGCGTCATCGAAGGCAC
Phenotype
MIM: 614724 MIM: 605739
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP63 PubMed Links

Gene Details

Gene
CEP63
Gene Name
centrosomal protein 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042383.1 9222 Intron NP_001035842.1
NM_001042384.1 9222 Intron NP_001035843.1
NM_001042400.1 9222 Intron NP_001035859.1
NM_025180.3 9222 Intron NP_079456.2
XM_005247795.4 9222 Intron XP_005247852.1
XM_005247797.3 9222 Intron XP_005247854.1
XM_005247799.3 9222 Intron XP_005247856.1
XM_006713760.3 9222 Intron XP_006713823.1
XM_011513194.2 9222 Intron XP_011511496.1
XM_011513197.2 9222 Intron XP_011511499.1
XM_017007244.1 9222 Intron XP_016862733.1
XM_017007245.1 9222 Intron XP_016862734.1
XM_017007246.1 9222 Intron XP_016862735.1
XM_017007247.1 9222 Intron XP_016862736.1
XM_017007248.1 9222 Intron XP_016862737.1
XM_017007249.1 9222 Intron XP_016862738.1
XM_017007250.1 9222 Intron XP_016862739.1
XM_017007251.1 9222 Intron XP_016862740.1
XM_017007252.1 9222 Intron XP_016862741.1
XM_017007253.1 9222 Intron XP_016862742.1
XM_017007254.1 9222 Intron XP_016862743.1
XM_017007255.1 9222 Intron XP_016862744.1
XM_017007256.1 9222 Intron XP_016862745.1
XM_017007257.1 9222 Intron XP_016862746.1
XM_017007258.1 9222 Intron XP_016862747.1
XM_017007259.1 9222 Intron XP_016862748.1
XM_017007260.1 9222 Intron XP_016862749.1
XM_017007261.1 9222 Intron XP_016862750.1
XM_017007262.1 9222 Intron XP_016862751.1
XM_017007263.1 9222 Intron XP_016862752.1
XM_017007264.1 9222 Intron XP_016862753.1
XM_017007265.1 9222 Intron XP_016862754.1
XM_017007266.1 9222 Intron XP_016862755.1
XM_017007267.1 9222 Intron XP_016862756.1
XM_017007268.1 9222 Intron XP_016862757.1
XM_017007269.1 9222 Intron XP_016862758.1
Gene
KY
Gene Name
kyphoscoliosis peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178554.4 9222 UTR 3 NP_848649.3
XM_005247418.3 9222 UTR 3 XP_005247475.1
XM_006713612.3 9222 UTR 3 XP_006713675.1
XM_017006287.1 9222 UTR 3 XP_016861776.1
XM_017006288.1 9222 UTR 3 XP_016861777.1
XM_017006289.1 9222 UTR 3 XP_016861778.1
XM_017006290.1 9222 UTR 3 XP_016861779.1
Gene
LOC101927568
Gene Name
uncharacterized LOC101927568
There are no transcripts associated with this gene.

View Full Product Details