Product Details

SNP ID
rs146768426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38008359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGACTTTTGGCAGCTGCTGCCC[C/T]GAAATGACCTGAGGAAAGGCAGAGG
Phenotype
MIM: 602142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLCD1 PubMed Links

Gene Details

Gene
PLCD1
Gene Name
phospholipase C delta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130964.1 2298 Silent Mutation TCA,TCG S658S NP_001124436.1
NM_006225.3 2298 Silent Mutation TCA,TCG S637S NP_006216.2
XM_017006622.1 2298 Silent Mutation TCA,TCG S579S XP_016862111.1
Gene
VILL
Gene Name
villin like
There are no transcripts associated with this gene.

View Full Product Details