Product Details

SNP ID
rs146791702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48454401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGAGCAAATAAACTGGCTGCTCC[C/T]TCTGTTTCCCATGTCAGGTAATGAT
Phenotype
MIM: 606605
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271022.1 837 Silent Mutation CCC,CCT P91P NP_001257951.1
NM_001271023.1 837 Silent Mutation CCC,CCT P125P NP_001257952.1
NM_032166.3 837 Silent Mutation CCC,CCT P218P NP_115542.2
NM_130384.2 837 Silent Mutation CCC,CCT P218P NP_569055.1
Gene
CCDC51
Gene Name
coiled-coil domain containing 51
There are no transcripts associated with this gene.

View Full Product Details