Product Details

SNP ID
rs146792880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49412496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGTGCTGGGCGCGCTGGGCAGC[A/G]AGTTCTTGCGGGAGTGGGAGGCGCA
Phenotype
MIM: 238310 MIM: 611516 MIM: 165390 MIM: 600690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMT PubMed Links

Gene Details

Gene
AMT
Gene Name
aminomethyltransferase
There are no transcripts associated with this gene.

Gene
NICN1
Gene Name
nicolin 1
There are no transcripts associated with this gene.

Gene
RHOA
Gene Name
ras homolog family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313941.1 291 Intron NP_001300870.1
NM_001313943.1 291 Intron NP_001300872.1
NM_001313944.1 291 Intron NP_001300873.1
NM_001313945.1 291 Intron NP_001300874.1
NM_001313946.1 291 Intron NP_001300875.1
NM_001313947.1 291 Intron NP_001300876.1
NM_001664.3 291 Intron NP_001655.1
Gene
TCTA
Gene Name
T-cell leukemia translocation altered
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022171.2 291 Missense Mutation AAG,GAG K24E NP_071503.1

View Full Product Details