Product Details

SNP ID
rs146902361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58566145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCCTTGAAGCTAAGCTCTGCTG[G/T]CTGCAGGTAAAACCACAGGTGGGGG
Phenotype
MIM: 608295
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM107A PubMed Links

Gene Details

Gene
FAM107A
Gene Name
family with sequence similarity 107 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076778.2 1587 UTR 3 NP_001070246.1
NM_001282713.1 1587 UTR 3 NP_001269642.1
NM_001282714.1 1587 UTR 3 NP_001269643.1
NM_007177.3 1587 UTR 3 NP_009108.1

View Full Product Details