Product Details

SNP ID
rs146922820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57557021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGCCCGAGGTCGGTGTCCCCTC[A/G]TCATTGTCTCCCTCCTCACCTTCTT
Phenotype
MIM: 616519
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE12 PubMed Links

Gene Details

Gene
PDE12
Gene Name
phosphodiesterase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322176.1 775 Silent Mutation TCA,TCG S214S NP_001309105.1
NM_001322177.1 775 Silent Mutation TCA,TCG S214S NP_001309106.1
NM_177966.6 775 Silent Mutation TCA,TCG S214S NP_808881.3

View Full Product Details