Product Details

SNP ID
rs146971607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49030617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTCAGGTGTCAGGTAAAAGGT[A/G]TCATTCCGGCCTTTCACACTCTGGG
Phenotype
MIM: 146691 MIM: 615150 MIM: 612911
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DALRD3 PubMed Links

Gene Details

Gene
DALRD3
Gene Name
DALR anticodon binding domain containing 3
There are no transcripts associated with this gene.

Gene
IMPDH2
Gene Name
inosine monophosphate dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000884.2 2584 Intron NP_000875.2
XM_006713128.3 2584 Intron XP_006713191.1
XM_017006349.1 2584 Intron XP_016861838.1
XM_017006350.1 2584 Intron XP_016861839.1
Gene
MIR191
Gene Name
microRNA 191
There are no transcripts associated with this gene.

Gene
NDUFAF3
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
There are no transcripts associated with this gene.

Gene
QRICH1
Gene Name
glutamine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320580.1 2584 Silent Mutation GAC,GAT D722D NP_001307509.1
NM_001320581.1 2584 Silent Mutation GAC,GAT D722D NP_001307510.1
NM_001320582.1 2584 Silent Mutation GAC,GAT D722D NP_001307511.1
NM_001320583.1 2584 Silent Mutation GAC,GAT D722D NP_001307512.1
NM_001320584.1 2584 Silent Mutation GAC,GAT D722D NP_001307513.1
NM_001320585.1 2584 Silent Mutation GAC,GAT D722D NP_001307514.1
NM_017730.3 2584 Silent Mutation GAC,GAT D722D NP_060200.2
NM_198880.2 2584 Silent Mutation GAC,GAT D722D NP_942581.1
XM_011533863.1 2584 Silent Mutation GAC,GAT D722D XP_011532165.1

View Full Product Details