Product Details

SNP ID
rs147018918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:97822260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCTGGCACAGCTTCTCCCGGTT[A/C]TTCGCTCCCCGAAGTCCTTCCCGGG
Phenotype
MIM: 608845
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARL6 PubMed Links

Gene Details

Gene
ARL6
Gene Name
ADP ribosylation factor like GTPase 6
There are no transcripts associated with this gene.

Gene
CRYBG3
Gene Name
crystallin beta-gamma domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153605.3 221 Missense Mutation TTA,TTC L18F NP_705833.3
XM_005247117.4 221 Intron XP_005247174.1

View Full Product Details