Product Details

SNP ID
rs147065790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:3150886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTTACAAGCAAAGTATTACTTT[A/G]TCTGGACTTATTTCATCTTCAGTGT
Phenotype
MIM: 609262 MIM: 612907
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRBN PubMed Links

Gene Details

Gene
CRBN
Gene Name
cereblon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173482.1 1335 Silent Mutation GAC,GAT D435D NP_001166953.1
NM_016302.3 1335 Silent Mutation GAC,GAT D436D NP_057386.2
XM_005265202.3 1335 Silent Mutation GAC,GAT D373D XP_005265259.1
XM_011533791.2 1335 Silent Mutation GAC,GAT D392D XP_011532093.1
XM_011533793.2 1335 Silent Mutation GAC,GAT D272D XP_011532095.1
XM_011533794.2 1335 Silent Mutation GAC,GAT D272D XP_011532096.1
Gene
TRNT1
Gene Name
tRNA nucleotidyl transferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302946.1 1335 Intron NP_001289875.1
NM_182916.2 1335 Intron NP_886552.2
XM_011533776.2 1335 Intron XP_011532078.1
XM_011533777.2 1335 Intron XP_011532079.1
XM_011533778.2 1335 UTR 3 XP_011532080.1

View Full Product Details