Product Details
- SNP ID
-
rs147239672
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:195279356 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATTTCTGTGAATCTTGCTGGAAA[C/T]GATTTAGTTTCTCTGGATTATTGGA
- Phenotype
-
MIM: 607766
MIM: 614552
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACAP2
PubMed Links
Gene Details
- Gene
- ACAP2
- Gene Name
- ArfGAP with coiled-coil, ankyrin repeat and PH domains 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012287.5 |
2676 |
Missense Mutation |
CAT,CGT |
H770R |
NP_036419.3 |
XM_006713557.3 |
2676 |
Missense Mutation |
CAT,CGT |
H777R |
XP_006713620.1 |
XM_011512602.2 |
2676 |
Missense Mutation |
CAT,CGT |
H812R |
XP_011510904.1 |
XM_011512603.2 |
2676 |
Missense Mutation |
CAT,CGT |
H805R |
XP_011510905.1 |
XM_011512604.2 |
2676 |
Missense Mutation |
CAT,CGT |
H768R |
XP_011510906.1 |
XM_011512605.2 |
2676 |
Missense Mutation |
CAT,CGT |
H768R |
XP_011510907.1 |
XM_011512606.2 |
2676 |
Missense Mutation |
CAT,CGT |
H768R |
XP_011510908.1 |
XM_017006046.1 |
2676 |
Missense Mutation |
CAT,CGT |
H840R |
XP_016861535.1 |
XM_017006047.1 |
2676 |
Missense Mutation |
CAT,CGT |
H833R |
XP_016861536.1 |
XM_017006048.1 |
2676 |
Missense Mutation |
CAT,CGT |
H805R |
XP_016861537.1 |
XM_017006049.1 |
2676 |
Missense Mutation |
CAT,CGT |
H798R |
XP_016861538.1 |
- Gene
- XXYLT1
- Gene Name
- xyloside xylosyltransferase 1
There are no transcripts associated with this gene.
View Full Product Details