Product Details

SNP ID
rs147245558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38697697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCATCGGAGAGTGGTTGCTAT[C/T]GGTTCATAGGATGATTTTGAAAGAT
Phenotype
MIM: 604427
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCN10A PubMed Links

Gene Details

Gene
SCN10A
Gene Name
sodium voltage-gated channel alpha subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293306.2 5229 Silent Mutation CCA,CCG P1840P NP_001280235.2
NM_001293307.2 5229 Silent Mutation CCA,CCG P1743P NP_001280236.2
NM_006514.3 5229 Silent Mutation CCA,CCG P1841P NP_006505.3
XM_005265371.3 5229 Silent Mutation CCA,CCG P1844P XP_005265428.1
XM_011533993.2 5229 Silent Mutation CCA,CCG P1843P XP_011532295.1
XM_011533994.2 5229 Silent Mutation CCA,CCG P1746P XP_011532296.1

View Full Product Details