Product Details
- SNP ID
-
rs147519719
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:50560957 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCACAGAAGTAGCAGCCTGTACA[C/G]AGGCGGCGTCCCGCCCATGCTCCAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
C3orf18
PubMed Links
Gene Details
- Gene
- C3orf18
- Gene Name
- chromosome 3 open reading frame 18
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001171740.2 |
937 |
Missense Mutation |
TCT,TGT |
S123C |
NP_001165211.1 |
NM_001171741.2 |
937 |
Missense Mutation |
TCT,TGT |
S123C |
NP_001165212.1 |
NM_001171743.2 |
937 |
Missense Mutation |
TCT,TGT |
S103C |
NP_001165214.2 |
NM_016210.4 |
937 |
Missense Mutation |
TCT,TGT |
S123C |
NP_057294.2 |
XM_011533781.2 |
937 |
Missense Mutation |
TCT,TGT |
S197C |
XP_011532083.1 |
XM_011533782.2 |
937 |
Missense Mutation |
TCT,TGT |
S127C |
XP_011532084.1 |
XM_011533783.2 |
937 |
Missense Mutation |
TCT,TGT |
S127C |
XP_011532085.1 |
XM_011533784.2 |
937 |
Missense Mutation |
TCT,TGT |
S127C |
XP_011532086.1 |
XM_011533785.2 |
937 |
Missense Mutation |
TCT,TGT |
S127C |
XP_011532087.1 |
XM_011533788.1 |
937 |
UTR 5 |
|
|
XP_011532090.1 |
XM_011533789.1 |
937 |
Intron |
|
|
XP_011532091.1 |
XM_011533790.1 |
937 |
Missense Mutation |
TCT,TGT |
S103C |
XP_011532092.1 |
XM_017006545.1 |
937 |
Intron |
|
|
XP_016862034.1 |
XM_017006546.1 |
937 |
Intron |
|
|
XP_016862035.1 |
XM_017006547.1 |
937 |
Missense Mutation |
TCT,TGT |
S103C |
XP_016862036.1 |
- Gene
- HEMK1
- Gene Name
- HemK methyltransferase family member 1
There are no transcripts associated with this gene.
View Full Product Details