Product Details
- SNP ID
-
rs147968046
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:51674333 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGGACCTGCTACTACTGGGCCTGA[C/T]TGGGGGCCTGACTCTCTTACTGCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RAD54L2
PubMed Links
Gene Details
- Gene
- RAD54L2
- Gene Name
- RAD54-like 2 (S. cerevisiae)
There are no transcripts associated with this gene.
- Gene
- TEX264
- Gene Name
- testis expressed 264
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001129884.2 |
337 |
Missense Mutation |
ACT,ATT |
T10I |
NP_001123356.1 |
NM_001243725.1 |
337 |
Missense Mutation |
ACT,ATT |
T10I |
NP_001230654.1 |
NM_001243726.1 |
337 |
Missense Mutation |
ACT,ATT |
T10I |
NP_001230655.1 |
NM_001243727.2 |
337 |
Missense Mutation |
ACT,ATT |
T10I |
NP_001230656.1 |
NM_001278195.1 |
337 |
Missense Mutation |
ACT,ATT |
T10I |
NP_001265124.1 |
NM_015926.5 |
337 |
Missense Mutation |
ACT,ATT |
T10I |
NP_057010.1 |
XM_006713195.3 |
337 |
Missense Mutation |
ACT,ATT |
T10I |
XP_006713258.1 |
XM_006713197.3 |
337 |
Missense Mutation |
ACT,ATT |
T10I |
XP_006713260.1 |
XM_006713198.2 |
337 |
UTR 5 |
|
|
XP_006713261.1 |
XM_011533805.2 |
337 |
Missense Mutation |
ACT,ATT |
T10I |
XP_011532107.1 |
XM_017006573.1 |
337 |
Missense Mutation |
ACT,ATT |
T49I |
XP_016862062.1 |
XM_017006574.1 |
337 |
Missense Mutation |
ACT,ATT |
T10I |
XP_016862063.1 |
View Full Product Details