Product Details
- SNP ID
-
rs148042871
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:58637187 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGCCACCAGCACCAGTGCACCCC[C/T]CGGAATTTCTTTAAGGAATTTCACT
- Phenotype
-
MIM: 608295
MIM: 608619
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM107A
PubMed Links
Gene Details
- Gene
- FAM107A
- Gene Name
- family with sequence similarity 107 member A
There are no transcripts associated with this gene.
- Gene
- FAM3D
- Gene Name
- family with sequence similarity 3 member D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138805.2 |
709 |
Missense Mutation |
AGG,GGG |
R138G |
NP_620160.1 |
XM_005264862.4 |
709 |
Missense Mutation |
AGG,GGG |
R138G |
XP_005264919.1 |
XM_005264863.4 |
709 |
Missense Mutation |
AGG,GGG |
R137G |
XP_005264920.1 |
XM_005264864.4 |
709 |
Missense Mutation |
AGG,GGG |
R137G |
XP_005264921.1 |
XM_006712965.3 |
709 |
Missense Mutation |
AGG,GGG |
R138G |
XP_006713028.1 |
XM_006712967.3 |
709 |
Missense Mutation |
AGG,GGG |
R101G |
XP_006713030.1 |
XM_011533348.2 |
709 |
Missense Mutation |
AGG,GGG |
R144G |
XP_011531650.1 |
XM_011533349.2 |
709 |
Missense Mutation |
AGG,GGG |
R144G |
XP_011531651.1 |
XM_011533350.2 |
709 |
Missense Mutation |
AGG,GGG |
R143G |
XP_011531652.1 |
XM_011533351.2 |
709 |
Missense Mutation |
AGG,GGG |
R138G |
XP_011531653.1 |
XM_011533352.2 |
709 |
Missense Mutation |
AGG,GGG |
R102G |
XP_011531654.1 |
XM_011533353.2 |
709 |
Missense Mutation |
AGG,GGG |
R102G |
XP_011531655.1 |
XM_011533354.2 |
709 |
Missense Mutation |
AGG,GGG |
R101G |
XP_011531656.1 |
- Gene
- FAM3D-AS1
- Gene Name
- FAM3D antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details