Product Details

SNP ID
rs148079551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50365419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGTTTACCGAGCCGAAGTAGTAC[C/T]GGGTCTGTTTCATGACGCAGCTGCT
Phenotype
MIM: 607082 MIM: 607068 MIM: 607069
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNA2D2 PubMed Links

Gene Details

Gene
CACNA2D2
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005505.2 3005 Missense Mutation CAG,CGG Q1012R NP_001005505.1
NM_001174051.2 3005 Missense Mutation CAG,CGG Q1019R NP_001167522.1
NM_001291101.1 3005 Missense Mutation CAG,CGG Q943R NP_001278030.1
NM_006030.3 3005 Missense Mutation CAG,CGG Q1012R NP_006021.2
XM_005265581.4 3005 Missense Mutation CAG,CGG Q1013R XP_005265638.1
XM_011534243.2 3005 Missense Mutation CAG,CGG Q1020R XP_011532545.1
Gene
CYB561D2
Gene Name
cytochrome b561 family member D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291284.1 3005 Intron NP_001278213.1
NM_007022.4 3005 Intron NP_008953.1
Gene
TMEM115
Gene Name
transmembrane protein 115
There are no transcripts associated with this gene.

View Full Product Details