Product Details
- SNP ID
-
rs148080961
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:70958286 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAATACTGCTGCGTGGAATGAATC[A/G]GCATTGTTCCTAGAGTTTGTCTCTC
- Phenotype
-
MIM: 605515
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FOXP1
PubMed Links
Gene Details
- Gene
- FOXP1
- Gene Name
- forkhead box P1
View Full Product Details