Product Details

SNP ID
rs148174212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48447062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCTCGCGTCACAGGCCCTGAGC[C/T]AATGTCCGGCCGCGGCTCGGGACGT
Phenotype
MIM: 606605 MIM: 615808
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271022.1 353 Intron NP_001257951.1
NM_001271023.1 353 Intron NP_001257952.1
NM_032166.3 353 Nonsense Mutation CAA,TAA Q73* NP_115542.2
NM_130384.2 353 Nonsense Mutation CAA,TAA Q73* NP_569055.1
Gene
CCDC51
Gene Name
coiled-coil domain containing 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256964.1 353 Intron NP_001243893.1
NM_001256965.2 353 Intron NP_001243894.1
NM_001256966.2 353 Intron NP_001243895.1
NM_001256967.2 353 Intron NP_001243896.1
NM_001256968.2 353 Intron NP_001243897.1
NM_001256969.2 353 Intron NP_001243898.1
NM_024661.4 353 Intron NP_078937.3
XM_011534113.2 353 Intron XP_011532415.1
Gene
TMA7
Gene Name
translation machinery associated 7 homolog
There are no transcripts associated with this gene.

View Full Product Details