Product Details

SNP ID
rs148290884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122190251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTAGAGCCCTTATGTGCTAAGT[A/G]CTTGTCGATTAAGAGATTCGGGGAA
Phenotype
MIM: 601199
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASR PubMed Links

Gene Details

Gene
CASR
Gene Name
calcium sensing receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000388.3 Intron NP_000379.2
NM_001178065.1 Intron NP_001171536.1
XM_005247837.2 Intron XP_005247894.1
XM_006713789.3 Intron XP_006713852.1
XM_017007324.1 Intron XP_016862813.1
XM_017007325.1 Intron XP_016862814.1

View Full Product Details