Product Details

SNP ID
rs148366584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49280803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGCAATGAGGTCGTACACATAA[A/G]GCCTTGCTGACAGGTTACAGACAAA
Phenotype
MIM: 603486
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf62 PubMed Links

Gene Details

Gene
C3orf62
Gene Name
chromosome 3 open reading frame 62
There are no transcripts associated with this gene.

Gene
MIR4271
Gene Name
microRNA 4271
There are no transcripts associated with this gene.

Gene
USP4
Gene Name
ubiquitin specific peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251877.1 2523 Intron NP_001238806.1
NM_003363.3 2523 Missense Mutation CCT,CTT P862L NP_003354.2
NM_199443.2 2523 Missense Mutation CCT,CTT P815L NP_955475.1

View Full Product Details