Product Details

SNP ID
rs148593044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186666103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGTGCCGTGAGTCCCACTGACTG[A/C]AGTGCTGTTGAGCCGGAGGCTGAGA
Phenotype
MIM: 142640
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HRG PubMed Links

Gene Details

Gene
HRG
Gene Name
histidine rich glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000412.4 152 Nonsense Mutation TGA,TGC *24C NP_000403.1
XM_005247415.3 152 Nonsense Mutation TGA,TGC *24C XP_005247472.1

View Full Product Details