Product Details

SNP ID
rs148657189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:36386895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTCCACGTGCACTTTGGAATCA[C/T]CTGATCAATTTGCACAAAAACACCT
Phenotype
MIM: 602317
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STAC PubMed Links

Gene Details

Gene
STAC
Gene Name
SH3 and cysteine rich domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292049.1 Intron NP_001278978.1
NM_003149.2 Intron NP_003140.1
XM_011534036.2 Intron XP_011532338.1
XM_011534037.2 Intron XP_011532339.1
XM_011534038.2 Intron XP_011532340.1
XM_017007083.1 Intron XP_016862572.1
XM_017007084.1 Intron XP_016862573.1

View Full Product Details