Product Details
- SNP ID
-
rs148752175
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:62399518 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCACAGTGAGACGGTTCCGGATC[A/G]TTTCATAGGTCTTGCTGTTTAAGGT
- Phenotype
-
MIM: 604667
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CADPS
PubMed Links
Gene Details
- Gene
- CADPS
- Gene Name
- calcium dependent secretion activator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003716.3 |
4491 |
Missense Mutation |
ACG,ATG |
T1317M |
NP_003707.2 |
NM_183393.2 |
4491 |
Missense Mutation |
ACG,ATG |
T1238M |
NP_899630.1 |
NM_183394.2 |
4491 |
Missense Mutation |
ACG,ATG |
T1278M |
NP_899631.1 |
XM_006713378.2 |
4491 |
Missense Mutation |
ACG,ATG |
T1259M |
XP_006713441.2 |
XM_011534177.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1395M |
XP_011532479.1 |
XM_011534178.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1389M |
XP_011532480.1 |
XM_011534180.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1379M |
XP_011532482.1 |
XM_011534182.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1372M |
XP_011532484.1 |
XM_011534183.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1366M |
XP_011532485.1 |
XM_011534189.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1340M |
XP_011532491.1 |
XM_011534191.2 |
4491 |
Missense Mutation |
ACG,ATG |
T1327M |
XP_011532493.1 |
XM_011534192.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1326M |
XP_011532494.1 |
XM_011534193.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1322M |
XP_011532495.1 |
XM_011534194.2 |
4491 |
Missense Mutation |
ACG,ATG |
T1321M |
XP_011532496.1 |
XM_011534195.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1317M |
XP_011532497.1 |
XM_011534196.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1316M |
XP_011532498.1 |
XM_011534197.2 |
4491 |
Missense Mutation |
ACG,ATG |
T1308M |
XP_011532499.1 |
XM_011534198.2 |
4491 |
Missense Mutation |
ACG,ATG |
T1261M |
XP_011532500.1 |
XM_011534199.2 |
4491 |
Missense Mutation |
ACG,ATG |
T1260M |
XP_011532501.1 |
XM_011534200.2 |
4491 |
Missense Mutation |
ACG,ATG |
T1255M |
XP_011532502.1 |
XM_011534202.2 |
4491 |
Intron |
|
|
XP_011532504.1 |
XM_011534203.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1311M |
XP_011532505.1 |
XM_017007358.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1397M |
XP_016862847.1 |
XM_017007359.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1393M |
XP_016862848.1 |
XM_017007360.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1390M |
XP_016862849.1 |
XM_017007361.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1374M |
XP_016862850.1 |
XM_017007362.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1370M |
XP_016862851.1 |
XM_017007363.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1367M |
XP_016862852.1 |
XM_017007364.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1341M |
XP_016862853.1 |
XM_017007365.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1337M |
XP_016862854.1 |
XM_017007366.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1335M |
XP_016862855.1 |
XM_017007367.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1333M |
XP_016862856.1 |
XM_017007368.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1332M |
XP_016862857.1 |
XM_017007369.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1326M |
XP_016862858.1 |
XM_017007370.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1321M |
XP_016862859.1 |
XM_017007371.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1318M |
XP_016862860.1 |
XM_017007372.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1318M |
XP_016862861.1 |
XM_017007373.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1317M |
XP_016862862.1 |
XM_017007374.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1314M |
XP_016862863.1 |
XM_017007375.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1313M |
XP_016862864.1 |
XM_017007376.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1312M |
XP_016862865.1 |
XM_017007377.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1310M |
XP_016862866.1 |
XM_017007378.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1309M |
XP_016862867.1 |
XM_017007379.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1308M |
XP_016862868.1 |
XM_017007380.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1289M |
XP_016862869.1 |
XM_017007381.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1288M |
XP_016862870.1 |
XM_017007382.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1284M |
XP_016862871.1 |
XM_017007383.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1282M |
XP_016862872.1 |
XM_017007384.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1272M |
XP_016862873.1 |
XM_017007385.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1268M |
XP_016862874.1 |
XM_017007386.1 |
4491 |
Missense Mutation |
ACG,ATG |
T1265M |
XP_016862875.1 |
View Full Product Details