Product Details

SNP ID
rs148994019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38138889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTGAGTACTTGGAGATCCGGCA[A/G]CTGGAGACACAAGCGGACCCCACTG
Phenotype
MIM: 604054 MIM: 602170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAA1 PubMed Links

Gene Details

Gene
ACAA1
Gene Name
acetyl-CoA acyltransferase 1
There are no transcripts associated with this gene.

Gene
MYD88
Gene Name
myeloid differentiation primary response 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172566.1 412 Silent Mutation CAA,CAG Q76Q NP_001166037.1
NM_001172567.1 412 Silent Mutation CAA,CAG Q76Q NP_001166038.1
NM_001172568.1 412 Silent Mutation CAA,CAG Q76Q NP_001166039.1
NM_001172569.1 412 Silent Mutation CAA,CAG Q76Q NP_001166040.1
NM_002468.4 412 Silent Mutation CAA,CAG Q76Q NP_002459.2
XM_005265172.1 412 Silent Mutation CAA,CAG Q76Q XP_005265229.1
XM_006713170.1 412 Silent Mutation CAA,CAG Q76Q XP_006713233.1

View Full Product Details