Product Details

SNP ID
rs149087483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49283691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGGTGAGGCTACCTGTGACAGA[C/T]GATGGACAGGAAGGCCAGTTAGGTG
Phenotype
MIM: 603486
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf62 PubMed Links

Gene Details

Gene
C3orf62
Gene Name
chromosome 3 open reading frame 62
There are no transcripts associated with this gene.

Gene
MIR4271
Gene Name
microRNA 4271
There are no transcripts associated with this gene.

Gene
USP4
Gene Name
ubiquitin specific peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251877.1 Intron NP_001238806.1
NM_003363.3 Intron NP_003354.2
NM_199443.2 Intron NP_955475.1

View Full Product Details