Product Details

SNP ID
rs149103210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69055479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGAATAGTACAGTCTGTGGGGT[A/G]GTGACATCAGCAACCGCCAGTTCTT
Phenotype
MIM: 601126 MIM: 603172
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR3136 PubMed Links

Gene Details

Gene
MIR3136
Gene Name
microRNA 3136
There are no transcripts associated with this gene.

Gene
TMF1
Gene Name
TATA element modulatory factor 1
There are no transcripts associated with this gene.

Gene
UBA3
Gene Name
ubiquitin like modifier activating enzyme 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003968.3 1284 Silent Mutation ACC,ACT T450T NP_003959.3
NM_198195.1 1284 Silent Mutation ACC,ACT T436T NP_937838.1
XM_011534210.1 1284 Silent Mutation ACC,ACT T423T XP_011532512.1
XM_011534211.1 1284 Silent Mutation ACC,ACT T409T XP_011532513.1

View Full Product Details