Product Details

SNP ID
rs149207587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138197697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCTTGCATGTGCTTTCCAGAGA[C/T]AGTTCTTGCTCCCTTGGACTTTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARMC8 PubMed Links

Gene Details

Gene
ARMC8
Gene Name
armadillo repeat containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267041.2 Intron NP_001253970.1
NM_001267042.2 Intron NP_001253971.1
NM_001282342.1 Intron NP_001269271.1
NM_014154.4 Intron NP_054873.2
NM_015396.5 Intron NP_056211.2
NM_213654.3 Intron NP_998819.1
XM_006713564.3 Intron XP_006713627.1
XM_006713565.2 Intron XP_006713628.1
XM_006713566.3 Intron XP_006713629.1
XM_006713567.3 Intron XP_006713630.1
XM_011512632.1 Intron XP_011510934.1
XM_011512633.1 Intron XP_011510935.1
XM_017006095.1 Intron XP_016861584.1
XM_017006096.1 Intron XP_016861585.1
XM_017006097.1 Intron XP_016861586.1
XM_017006098.1 Intron XP_016861587.1
XM_017006099.1 Intron XP_016861588.1
XM_017006100.1 Intron XP_016861589.1
XM_017006101.1 Intron XP_016861590.1

View Full Product Details