Product Details

SNP ID
rs149215876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:74264157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTTAAACTCATAGTTTAAGAAC[A/C]TTTATATCAAAAGTCTGAAGTATGT
Phenotype
MIM: 601325
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNTN3 PubMed Links

Gene Details

Gene
CNTN3
Gene Name
contactin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020872.2 2038 UTR 3 NP_065923.1
XM_005264757.3 2038 UTR 3 XP_005264814.1
XM_011533768.2 2038 UTR 3 XP_011532070.1
XM_017006507.1 2038 UTR 3 XP_016861996.1
XM_017006508.1 2038 UTR 3 XP_016861997.1
XM_017006509.1 2038 UTR 3 XP_016861998.1

View Full Product Details