Product Details
- SNP ID
-
rs149263533
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:184036512 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCTTCCATGGACAGAGGTGAAC[A/G]CTCTCCTTCAGCCCTTTCACCTACA
- Phenotype
-
MIM: 610122
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HTR3D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6443930] are located under a probe and SNP(s) [rs73183412] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HTR3D
- Gene Name
- 5-hydroxytryptamine receptor 3D
View Full Product Details