Product Details

SNP ID
rs149264259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53864798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCACACTACTGCCCCCCATTAA[C/G]GTTCTTGTGGTTTACCCATCTGAAA
Phenotype
MIM: 605458
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACTR8 PubMed Links

Gene Details

Gene
ACTR8
Gene Name
ARP8 actin-related protein 8 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022899.4 907 Intron NP_075050.3
XM_005265587.4 907 UTR 3 XP_005265644.1
XM_011534249.2 907 Intron XP_011532551.1
Gene
IL17RB
Gene Name
interleukin 17 receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018725.3 907 Missense Mutation AAC,AAG N333K NP_061195.2
XM_005265310.4 907 Missense Mutation AAC,AAG N362K XP_005265367.1
XM_005265311.4 907 Missense Mutation AAC,AAG N346K XP_005265368.1
XM_005265312.4 907 Missense Mutation AAC,AAG N317K XP_005265369.1
XM_011533941.2 907 Intron XP_011532243.1
XM_017006804.1 907 Missense Mutation AAC,AAG N245K XP_016862293.1
XM_017006805.1 907 Missense Mutation AAC,AAG N229K XP_016862294.1
XM_017006806.1 907 Missense Mutation AAC,AAG N216K XP_016862295.1
XM_017006807.1 907 Intron XP_016862296.1

View Full Product Details