Product Details
- SNP ID
-
rs149482869
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:37055116 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCCAAGGTAGTTATATCCTGTTC[C/T]GCTTTTGAAAGCTTAAATTTGTATT
- Phenotype
-
MIM: 614043
MIM: 120436
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LRRFIP2
PubMed Links
Gene Details
- Gene
- LRRFIP2
- Gene Name
- LRR binding FLII interacting protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001134369.2 |
1716 |
Silent Mutation |
GCA,GCG |
A343A |
NP_001127841.1 |
NM_001282691.1 |
1716 |
Silent Mutation |
GCA,GCG |
A422A |
NP_001269620.1 |
NM_006309.3 |
1716 |
Silent Mutation |
GCA,GCG |
A640A |
NP_006300.1 |
NM_017724.2 |
1716 |
Silent Mutation |
GCA,GCG |
A319A |
NP_060194.1 |
XM_005265539.2 |
1716 |
Silent Mutation |
GCA,GCG |
A640A |
XP_005265596.1 |
XM_005265540.1 |
1716 |
Silent Mutation |
GCA,GCG |
A640A |
XP_005265597.1 |
XM_005265550.1 |
1716 |
Silent Mutation |
GCA,GCG |
A496A |
XP_005265607.1 |
XM_005265551.1 |
1716 |
Silent Mutation |
GCA,GCG |
A459A |
XP_005265608.1 |
XM_005265553.1 |
1716 |
Silent Mutation |
GCA,GCG |
A408A |
XP_005265610.1 |
XM_005265554.2 |
1716 |
Silent Mutation |
GCA,GCG |
A408A |
XP_005265611.1 |
XM_005265556.1 |
1716 |
Silent Mutation |
GCA,GCG |
A384A |
XP_005265613.1 |
XM_006713385.1 |
1716 |
Silent Mutation |
GCA,GCG |
A639A |
XP_006713448.1 |
XM_006713386.1 |
1716 |
Silent Mutation |
GCA,GCG |
A626A |
XP_006713449.1 |
XM_006713387.1 |
1716 |
Silent Mutation |
GCA,GCG |
A625A |
XP_006713450.1 |
XM_006713388.1 |
1716 |
Silent Mutation |
GCA,GCG |
A624A |
XP_006713451.1 |
XM_006713389.1 |
1716 |
Silent Mutation |
GCA,GCG |
A623A |
XP_006713452.1 |
XM_006713390.1 |
1716 |
Silent Mutation |
GCA,GCG |
A623A |
XP_006713453.1 |
XM_006713392.1 |
1716 |
Silent Mutation |
GCA,GCG |
A616A |
XP_006713455.1 |
XM_006713393.1 |
1716 |
Silent Mutation |
GCA,GCG |
A610A |
XP_006713456.1 |
XM_006713394.1 |
1716 |
Silent Mutation |
GCA,GCG |
A609A |
XP_006713457.1 |
XM_006713395.1 |
1716 |
Silent Mutation |
GCA,GCG |
A609A |
XP_006713458.1 |
XM_006713396.1 |
1716 |
Silent Mutation |
GCA,GCG |
A604A |
XP_006713459.1 |
XM_006713397.1 |
1716 |
Silent Mutation |
GCA,GCG |
A568A |
XP_006713460.1 |
XM_006713400.1 |
1716 |
Silent Mutation |
GCA,GCG |
A423A |
XP_006713463.1 |
XM_006713404.1 |
1716 |
Silent Mutation |
GCA,GCG |
A334A |
XP_006713467.1 |
XM_011534217.1 |
1716 |
Silent Mutation |
GCA,GCG |
A640A |
XP_011532519.1 |
XM_011534218.1 |
1716 |
Silent Mutation |
GCA,GCG |
A640A |
XP_011532520.1 |
XM_011534219.1 |
1716 |
Silent Mutation |
GCA,GCG |
A640A |
XP_011532521.1 |
XM_011534221.1 |
1716 |
Silent Mutation |
GCA,GCG |
A604A |
XP_011532523.1 |
XM_011534222.1 |
1716 |
Silent Mutation |
GCA,GCG |
A575A |
XP_011532524.1 |
XM_011534229.1 |
1716 |
Silent Mutation |
GCA,GCG |
A423A |
XP_011532531.1 |
XM_017007465.1 |
1716 |
Silent Mutation |
GCA,GCG |
A589A |
XP_016862954.1 |
XM_017007466.1 |
1716 |
Silent Mutation |
GCA,GCG |
A587A |
XP_016862955.1 |
XM_017007467.1 |
1716 |
Silent Mutation |
GCA,GCG |
A585A |
XP_016862956.1 |
XM_017007468.1 |
1716 |
Silent Mutation |
GCA,GCG |
A553A |
XP_016862957.1 |
XM_017007469.1 |
1716 |
Silent Mutation |
GCA,GCG |
A551A |
XP_016862958.1 |
XM_017007470.1 |
1716 |
Silent Mutation |
GCA,GCG |
A539A |
XP_016862959.1 |
XM_017007471.1 |
1716 |
Silent Mutation |
GCA,GCG |
A442A |
XP_016862960.1 |
XM_017007472.1 |
1716 |
Silent Mutation |
GCA,GCG |
A428A |
XP_016862961.1 |
XM_017007473.1 |
1716 |
Silent Mutation |
GCA,GCG |
A418A |
XP_016862962.1 |
XM_017007474.1 |
1716 |
Silent Mutation |
GCA,GCG |
A411A |
XP_016862963.1 |
XM_017007475.1 |
1716 |
Silent Mutation |
GCA,GCG |
A411A |
XP_016862964.1 |
XM_017007476.1 |
1716 |
Silent Mutation |
GCA,GCG |
A408A |
XP_016862965.1 |
XM_017007477.1 |
1716 |
Silent Mutation |
GCA,GCG |
A387A |
XP_016862966.1 |
XM_017007478.1 |
1716 |
Silent Mutation |
GCA,GCG |
A377A |
XP_016862967.1 |
XM_017007479.1 |
1716 |
Silent Mutation |
GCA,GCG |
A377A |
XP_016862968.1 |
XM_017007480.1 |
1716 |
Silent Mutation |
GCA,GCG |
A377A |
XP_016862969.1 |
XM_017007481.1 |
1716 |
Silent Mutation |
GCA,GCG |
A374A |
XP_016862970.1 |
XM_017007482.1 |
1716 |
Silent Mutation |
GCA,GCG |
A353A |
XP_016862971.1 |
XM_017007483.1 |
1716 |
Silent Mutation |
GCA,GCG |
A343A |
XP_016862972.1 |
XM_017007484.1 |
1716 |
Silent Mutation |
GCA,GCG |
A343A |
XP_016862973.1 |
- Gene
- MLH1
- Gene Name
- mutL homolog 1
There are no transcripts associated with this gene.
View Full Product Details