Product Details

SNP ID
rs149834277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14490335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGAGCAAGGCCTGCTGCCTCCC[C/T]TGGCCTTCCCCCTGCCTACACCACT
Phenotype
MIM: 186854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRIP2 PubMed Links

Gene Details

Gene
GRIP2
Gene Name
glutamate receptor interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080423.3 6497 UTR 3 NP_001073892.3
XM_011534141.2 6497 Intron XP_011532443.1
XM_011534142.2 6497 Intron XP_011532444.1
XM_011534143.2 6497 Intron XP_011532445.1
XM_011534145.2 6497 Intron XP_011532447.1
XM_011534146.2 6497 Intron XP_011532448.1
Gene
SLC6A6
Gene Name
solute carrier family 6 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134367.3 6497 Intron NP_001127839.2
NM_001134368.3 6497 Intron NP_001127840.1
NM_003043.5 6497 Intron NP_003034.2
XM_006713307.2 6497 Intron XP_006713370.1
XM_011534030.2 6497 Intron XP_011532332.1
XM_011534031.2 6497 Intron XP_011532333.1

View Full Product Details