Product Details

SNP ID
rs149905428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40530270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGAGAGAGGGGAAGCACCATG[A/G]GGCCCAGATCCCTGGGACACCGAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF621 PubMed Links

Gene Details

Gene
ZNF621
Gene Name
zinc finger protein 621
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098414.2 499 Nonsense Mutation TGA,TGG *71W NP_001091884.1
NM_001287245.1 499 Nonsense Mutation TGA,TGG *71W NP_001274174.1
NM_198484.4 499 Nonsense Mutation TGA,TGG *71W NP_940886.1
XM_005265079.3 499 Nonsense Mutation TGA,TGG *63W XP_005265136.1
XM_005265080.3 499 Nonsense Mutation TGA,TGG *36W XP_005265137.1
XM_006713114.3 499 Nonsense Mutation TGA,TGG *63W XP_006713177.1
XM_011533610.2 499 Nonsense Mutation TGA,TGG *63W XP_011531912.1
XM_017006226.1 499 Nonsense Mutation TGA,TGG *36W XP_016861715.1

View Full Product Details