Product Details

SNP ID
rs149978587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98282906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGGGATGTCGAATGAGGACATG[C/G]AACAGGATAATACAACATTGCTGAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5H2 PubMed Links

Gene Details

Gene
OR5H2
Gene Name
olfactory receptor family 5 subfamily H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005482.1 19 Missense Mutation CAA,GAA Q7E NP_001005482.1

View Full Product Details