Product Details

SNP ID
rs149994975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126607171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAACTTTTGGAAATAATACTAT[C/T]AAGATGAACATGAAAAAGCAATTCA
Phenotype
MIM: 606235
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXNRD3 PubMed Links

Gene Details

Gene
TXNRD3
Gene Name
thioredoxin reductase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173513.1 2800 UTR 3 NP_001166984.1
NM_052883.1 2800 UTR 3 NP_443115.1
Gene
TXNRD3NB
Gene Name
thioredoxin reductase 3 neighbor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039783.1 2800 Intron NP_001034872.1

View Full Product Details